accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNNH3COHOOHOOHOOHO(E)(E)

CAS RN 87233-62-3

IUPAC Name: bis((2E)-but-2-enedioic acid); 1-(2-ethoxyethyl)-2-(4-methyl-1,4-diazepan-1-yl)-1H-1,3-benzodiazole
Synonyms: emedastine; bis(fumaric acid)
Molecular Weight (g/mol): 534.57
Molecular Formula: C25H34N4O9
InChi Key: FWLKKPKZQYVAFR-LVEZLNDCSA-N
SMILES: OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.CCOCCN1C(=NC2=CC=CC=C12)N1CCCN(C)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results