Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Emedastine Difumarate 98.0+%, TCI America™
SDP

Catalog No. E0936200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0936200MG 200 mg
E09361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0936200MG Supplier TCI America Supplier No. E0936200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87233-62-3
Molecular Formula C25H34N4O9
Molecular Weight (g/mol) 534.566
MDL Number MFCD00923844
InChI Key FWLKKPKZQYVAFR-LVEZLNDCSA-N
Synonym 1-(2-Ethoxyethyl)-2-(4-methyl-1-homopiperazinyl)benzimidazole Difumarate
PubChem CID 5282485
IUPAC Name (E)-but-2-enedioic acid;1-(2-ethoxyethyl)-2-(4-methyl-1,4-diazepan-1-yl)benzimidazole
SMILES CCOCCN1C2=CC=CC=C2N=C1N3CCCN(CC3)C.C(=CC(=O)O)C(=O)O.C(=CC(=O)O)C(=O)O

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 200 mg
Formula Weight 534.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Emedastine Difumarate
RTECSNumber DD8870000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.