accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 872672-50-9

IUPAC Name: 1-ethyl-3-methylpyridin-1-ium ethyl sulfate
Synonyms: 1-ethyl-3-methylpyridin-1-ium ethyl sulfate
Molecular Weight (g/mol): 247.31
Molecular Formula: C10H17NO4S
InChi Key: ZTLWMUBOQHZKNS-UHFFFAOYSA-M
SMILES: CCOS([O-])(=O)=O.CC[N+]1=CC=CC(C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results