Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylpyridinium Ethyl Sulfate 98.0+%, TCI America™
SDP

Catalog No. E06815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06815G 5 g
E068125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06815G Supplier TCI America Supplier No. E06815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 872672-50-9
Molecular Formula C10H17NO4S
Molecular Weight (g/mol) 247.309
InChI Key ZTLWMUBOQHZKNS-UHFFFAOYSA-M
PubChem CID 44629805
IUPAC Name 1-ethyl-3-methylpyridin-1-ium;ethyl sulfate
SMILES CC[N+]1=CC=CC(=C1)C.CCOS(=O)(=O)[O-]

Specifications

Quantity 5 g
Formula Weight 247.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylpyridinium Ethyl Sulfate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.