accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 873-63-2

HOCl

CAS RN 873-63-2

IUPAC Name: (3-chlorophenyl)methanol
Synonyms: M-chlorobenzyl alcohol
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: ZSRDNPVYGSFUMD-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results