Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. C065010ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C065010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C065010ML Supplier TCI America Supplier No. C065010ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 873-63-2
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00004632
InChI Key ZSRDNPVYGSFUMD-UHFFFAOYSA-N
Synonym 3-chlorobenzyl alcohol, 3-chlorophenyl methanol, m-chlorobenzyl alcohol, benzenemethanol, 3-chloro, 3-chlorobenzylalcohol, 3-chlorophenyl methan-1-ol, m-chlorobenzylalcohol, 3-chlorobenyl alcohol, pubchem3626, acmc-209qjt
PubChem CID 70117
IUPAC Name (3-chlorophenyl)methanol
SMILES C1=CC(=CC(=C1)Cl)CO

Specifications

Color Yellow
Boiling Point 242°C
Quantity 10 mL
Formula Weight 142.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzyl Alcohol
EINECSNumber (3)-1050
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.