accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87314-14-5

IUPAC Name: λ³-nickel(3+) bis((3,6-dichloro-2-sulfanidylphenyl)sulfanide) tetrabutylazanium
Synonyms: λ³-nickel(3+) bis((3,6-dichloro-2-sulfanidylphenyl)sulfanide) tetrabutylammonium ion
Molecular Weight (g/mol): 719.37
Molecular Formula: C28H40Cl4NNiS4
InChi Key: DPTWRZYDNJMUTE-UHFFFAOYSA-J
SMILES: [Ni+3].[S-]C1=C(Cl)C=CC(Cl)=C1[S-].[S-]C1=C(Cl)C=CC(Cl)=C1[S-].CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results