Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Bis(3,6-dichloro-1,2-benzenedithiolato)nickelate 98.0+%, TCI America™
SDP

Catalog No. T3204200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3204200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3204200MG Supplier TCI America Supplier No. T3204200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87314-14-5
Molecular Formula C28H40Cl4NNiS4
Molecular Weight (g/mol) 719.37
MDL Number MFCD00274632
InChI Key DPTWRZYDNJMUTE-UHFFFAOYSA-J
Synonym Tetrabutylammonium Bis(3,6-dichloro-1,2-benzenedithiolato)nickel(III) Complex
PubChem CID 16212902
IUPAC Name λ³-nickel(3+) bis((3,6-dichloro-2-sulfanidylphenyl)sulfanide) tetrabutylazanium
SMILES [Ni+3].[S-]C1=C(Cl)C=CC(Cl)=C1[S-].[S-]C1=C(Cl)C=CC(Cl)=C1[S-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 205°C
Color Green
Quantity 200 mg
Formula Weight 719.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Bis(3,6-dichloro-1,2-benzenedithiolato)nickelate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.