CAS RN 873225-46-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ IKK Inhibitor VII
The IKK Inhibitor VII, also referenced under CAS 873225-46-8, controls the biological activity of IKK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.