accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87331-47-3

IUPAC Name: 5-(1,3-benzoxazol-2-yl)-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms: 5-(1,3-benzoxazol-2-yl)-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Molecular Weight (g/mol): 358.4
Molecular Formula: C22H18N2O3
InChi Key: XYFLALJXCJWBPG-UHFFFAOYSA-N
SMILES: O=C1OC2=C3CCCN4CCCC(C=C2C=C1C1=NC2=CC=CC=C2O1)=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results