Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 525 97.0+%, TCI America™
SDP

Catalog No. C3047200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3047200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3047200MG Supplier TCI America Supplier No. C3047200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87331-47-3
Molecular Formula C22H18N2O3
Molecular Weight (g/mol) 358.397
InChI Key XYFLALJXCJWBPG-UHFFFAOYSA-N
Synonym 10-(2-Benzoxazolyl)-2,3,6,7-tetrahydro-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one
PubChem CID 1371421
SMILES C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C5=NC6=CC=CC=C6O5)CCCN3C1

Specifications

Melting Point 278°C
Quantity 200 mg
Formula Weight 358.40
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.