accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONN

CAS RN 87349-92-6

IUPAC Name: 5-(pyridin-3-yl)-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms: 5-(pyridin-3-yl)-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Molecular Weight (g/mol): 318.38
Molecular Formula: C20H18N2O2
InChi Key: OEEAIIVRKIVRNX-UHFFFAOYSA-N
SMILES: O=C1OC2=C3CCCN4CCCC(C=C2C=C1C1=CN=CC=C1)=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results