Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coumarin 510 98.0+%, TCI America™
SDP

Catalog No. C3072200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3072200MG 200 mg
C30721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3072200MG Supplier TCI America Supplier No. C3072200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 87349-92-6
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.376
MDL Number MFCD00467042
InChI Key OEEAIIVRKIVRNX-UHFFFAOYSA-N
Synonym 2,3,6,7-Tetrahydro-10-(3-pyridyl)-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one
PubChem CID 9905078
SMILES C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C5=CN=CC=C5)CCCN3C1

Specifications

Melting Point 165°C
Color Yellow
Quantity 200 mg
Formula Weight 318.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 510
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.