accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFH3CNCH3NH3CCH3ONNNN

CAS RN 873798-09-5

IUPAC Name: [(dimethylamino)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})methylidene]dimethylazanium; tetrafluoroboranuide
Synonyms: [(dimethylamino)({[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})methylidene]dimethylazanium tetrafluoroborate
Molecular Weight (g/mol): 322.07
Molecular Formula: C10H15BF4N6O
InChi Key: AUPDFAPCZZXFMX-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results