accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOH

CAS RN 87392-05-0

IUPAC Name: oxolane-2-carboxylic acid
Synonyms: tetrahydro-2-furoic acid
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: UJJLJRQIPMGXEZ-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCO1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)