accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87394-63-6

IUPAC Name: 1-[3-(trifluoromethyl)pyridin-2-yl]piperazine
Synonyms: 1-[3-(trifluoromethyl)pyridin-2-yl]piperazine
Molecular Weight (g/mol): 231.22
Molecular Formula: C10H12F3N3
InChi Key: FRFKCMNQNNNZNO-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(N=CC=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results