Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[3-(Trifluoromethyl)-2-pyridyl]piperazine, ≥95%, Thermo Scientific™

Catalog No. SEW04032DE
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SEW04032DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW04032DE Supplier Thermo Scientific Chemicals Supplier No. SEW04032DE

missing translation for 'validMainframeMsg'

Specifications

CAS 87394-63-6
CAS Min % 95.0
Assay Percent Range 95%
Molecular Formula C10H12F3N3
Quantity 5g
Synonym 1-3-trifluoromethyl pyridin-2-yl piperazine, 1-3-trifluoromethyl pyrid-2-yl piperazine, 1-3-trifluoromethyl-pyridin-2-yl-piperazine, 1-3-trifluoromethyl-2-pyridyl piperazine, chembl45663, 1-3-trifluoromethylpyridin-2-yl piperazine, 1-3-trifluoromethyl-2-pyridinyl piperazine, 1-3-trifluoromethyl pyridine-2-yl piperazine, 3-trifluoromethyl-2-pyridyl piperazine
InChI Key FRFKCMNQNNNZNO-UHFFFAOYSA-N
SMILES C1CN(CCN1)C2=C(C=CC=N2)C(F)(F)F
IUPAC Name 1-[3-(trifluoromethyl)pyridin-2-yl]piperazine
Molecular Weight (g/mol) 231.222
PubChem CID 2777786
Formula Weight 231.22
Percent Purity 95%
Chemical Name or Material 1-[3-(Trifluoromethyl)-2-pyridyl]piperazine
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.