accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874-77-1

IUPAC Name: 2-(4-methylpiperazin-1-yl)acetonitrile
Synonyms: 2-(4-methylpiperazin-1-yl)acetonitrile
Molecular Weight (g/mol): 139.2
Molecular Formula: C7H13N3
InChi Key: XCUQQNRWWCWVOP-UHFFFAOYSA-N
SMILES: CN1CCN(CC#N)CC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results