accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874219-47-3

IUPAC Name: [5-(tert-butylsulfamoyl)-2-methylphenyl]boronic acid
Synonyms: 5-(tert-butylsulfamoyl)-2-methylphenylboronic acid
Molecular Weight (g/mol): 271.14
Molecular Formula: C11H18BNO4S
InChi Key: PDKNOWSWRNUQHQ-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1)S(=O)(=O)NC(C)(C)C)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results