Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-tert-Butylsulfamoyl-2-methylbenzeneboronic acid, 95%, Thermo Scientific™

Catalog No. AAH53316MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53316MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH53316MD Supplier Thermo Scientific Chemicals Supplier No. H53316MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874219-47-3
Molecular Formula C11H18BNO4S
Molecular Weight (g/mol) 271.138
MDL Number MFCD08235063
InChI Key PDKNOWSWRNUQHQ-UHFFFAOYSA-N
Synonym 5-n-tert-butyl sulfamoyl-2-methylphenyl boronic acid, n-tert-butyl 3-borono-4-methylbenzenesulfonamide, 5-n-tert-butylsulfamoyl-2-methylbenzeneboronic acid, 5-n-tert-butylsulphamoyl-2-methylbenzeneboronic acid, boronic acid,b-5-1,1-dimethylethyl amino sulfonyl-2-methylphenyl, 5-tert-butylsulfamoyl-2-methylphenylboronic acid, 5-tert-butylsulfamoyl-2-methylphenyl boronic acid, acmc-209qlb, n-tert-butyl-3-borono-4-methylbenzenesulfonamide, 5-tert-butylsulfamoyl-2-methyl-phenyl boronic acid
PubChem CID 44119818
IUPAC Name [5-(tert-butylsulfamoyl)-2-methylphenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)S(=O)(=O)NC(C)(C)C)C)(O)O

Specifications

Melting Point 142°C to 145°C
Quantity 250 mg
Formula Weight 271.14
Percent Purity 95%
Chemical Name or Material 5-tert-Butylsulfamoyl-2-methylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.