accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874288-38-7

H3COOFBOHHO

CAS RN 874288-38-7

IUPAC Name: [4-(ethoxycarbonyl)-3-fluorophenyl]boronic acid
Synonyms: 4-(ethoxycarbonyl)-3-fluorophenylboronic acid
Molecular Weight (g/mol): 211.98
Molecular Formula: C9H10BFO4
InChi Key: CXSPPZZTTSHTMP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1F)B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results