Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Ethoxycarbonyl)-3-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E12075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1207-5G 5 g
E12071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1207-5G Supplier TCI America Supplier No. E12075G
Add to Cart
Add to Cart

Specifications

CAS 874288-38-7
Melting Point 161°C
Color White
Molecular Formula C9H10BFO4
Quantity 5 g
MDL Number MFCD06656274
Synonym 4-ethoxycarbonyl-3-fluorophenylboronic acid, 4-ethoxycarbonyl-3-fluorobenzeneboronic acid, ethyl 4-borono-2-fluorobenzoate, 4-ethoxycarbonyl-3-fluorophenyl boronic acid, 4-ethoxycarbonyl-3-fluoro-phenyl boronic acid
InChI Key CXSPPZZTTSHTMP-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)C(=O)OCC)F)(O)O
IUPAC Name (4-ethoxycarbonyl-3-fluorophenyl)boronic acid
Molecular Weight (g/mol) 211.983
PubChem CID 2763245
Formula Weight 211.98
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.