accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874459-85-5

IUPAC Name: [2-(methylcarbamoyl)phenyl]boronic acid
Synonyms: 2-(methylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 178.98
Molecular Formula: C8H10BNO3
InChi Key: AXWDCOSDMJAMOB-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results