accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874814-95-6

IUPAC Name: 1-(4-fluoro-2-nitrophenyl)-1,4-diazepane
Synonyms: 1-(4-fluoro-2-nitrophenyl)-1,4-diazepane
Molecular Weight (g/mol): 239.25
Molecular Formula: C11H14FN3O2
InChi Key: HFLSWHSIIVSRPS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(F)=C1)N1CCCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results