accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 874815-01-7

IUPAC Name: 1-(2-chloro-6-nitrophenyl)-1,4-diazepane
Synonyms: 1-(2-chloro-6-nitrophenyl)-1,4-diazepane
Molecular Weight (g/mol): 255.7
Molecular Formula: C11H14ClN3O2
InChi Key: DGSFYWNASDEFDR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(N2CCCNCC2)C(Cl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results