accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFNOHNH2(E/Z)

CAS RN 874880-59-8

IUPAC Name: 3,5-difluoro-N'-hydroxybenzene-1-carboximidamide
Synonyms: 3,5-difluoro-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 172.14
Molecular Formula: C7H6F2N2O
InChi Key: GYTSCORJTXOMMI-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC(F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results