accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 87592-41-4

IUPAC Name: 4-(4-ethylcyclohexyl)benzoic acid
Synonyms: 4-(4-ethylcyclohexyl)benzoic acid
Molecular Weight (g/mol): 232.32
Molecular Formula: C15H20O2
InChi Key: DUJHWZDXZFNEKU-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results