Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(trans-4-Ethylcyclohexyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. E12535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E12535G Supplier TCI America Supplier No. E12535G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87592-41-4
Molecular Formula C15H20O2
Molecular Weight (g/mol) 232.323
MDL Number MFCD06658176
InChI Key DUJHWZDXZFNEKU-UHFFFAOYSA-N
PubChem CID 13955174
IUPAC Name 4-(4-ethylcyclohexyl)benzoic acid
SMILES CCC1CCC(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 232.32
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Ethylcyclohexyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.