accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SiH3CH3CCH3CH3N

CAS RN 87630-35-1

IUPAC Name: 1-[tris(propan-2-yl)silyl]-1H-pyrrole
Synonyms: 1-(triisopropylsilyl)pyrrole
Molecular Weight (g/mol): 223.44
Molecular Formula: C13H25NSi
InChi Key: FBQURXLBJJNDBX-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)(C(C)C)N1C=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)