accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNSOOCl

CAS RN 876316-36-8

IUPAC Name: 1-methyl-1H-indole-4-sulfonyl chloride
Synonyms: 1-methylindole-4-sulfonyl chloride
Molecular Weight (g/mol): 229.68
Molecular Formula: C9H8ClNO2S
InChi Key: HFWFOJFQXZBLAU-UHFFFAOYSA-N
SMILES: CN1C=CC2=C(C=CC=C12)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results