accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 876316-38-0

IUPAC Name: 1-[2-(pyrrolidin-1-yl)pyridin-4-yl]methanamine
Synonyms: 1-[2-(pyrrolidin-1-yl)pyridin-4-yl]methanamine
Molecular Weight (g/mol): 177.25
Molecular Formula: C10H15N3
InChi Key: BNVQNNKKUNFIND-UHFFFAOYSA-N
SMILES: NCC1=CC(=NC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results