accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 876316-41-5

IUPAC Name: 2-[(1H-1,2,4-triazol-1-yl)methyl]benzonitrile
Synonyms: 2-(1,2,4-triazol-1-ylmethyl)benzonitrile
Molecular Weight (g/mol): 184.2
Molecular Formula: C10H8N4
InChi Key: AFAFVWQAHQRISI-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1CN1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results