accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNClO

CAS RN 876316-48-2

IUPAC Name: 3-(chloromethyl)-5-(furan-2-yl)-1-methyl-1H-pyrazole
Synonyms: 3-(chloromethyl)-5-(furan-2-yl)-1-methylpyrazole
Molecular Weight (g/mol): 196.63
Molecular Formula: C9H9ClN2O
InChi Key: DZUIHVJDKDNADW-UHFFFAOYSA-N
SMILES: CN1N=C(CCl)C=C1C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results