Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Chloromethyl)-5-(2-furyl)-1-methyl-1H-pyrazole, 95%, Thermo Scientific™

Catalog No. CC46924EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC46924EA 10 g
CC46924CB 250 mg
CC46924DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC46924EA Supplier Thermo Scientific Chemicals Supplier No. CC46924EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 876316-48-2
Molecular Formula C9H9ClN2O
Molecular Weight (g/mol) 196.63
MDL Number MFCD08690269
InChI Key DZUIHVJDKDNADW-UHFFFAOYSA-N
Synonym 3-chloromethyl-5-2-furyl-1-methyl-1h-pyrazole, 3-chloromethyl-5-furan-2-yl-1-methylpyrazole, 1h-pyrazole,3-chloromethyl-5-2-furanyl-1-methyl, 3-chloromethyl-5-furan-2-yl-1-methyl-1h-pyrazole, 2-3-chloromethyl-1-methylpyrazol-5-yl furan
PubChem CID 18525803
IUPAC Name 3-(chloromethyl)-5-(furan-2-yl)-1-methylpyrazole
SMILES CN1N=C(CCl)C=C1C1=CC=CO1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 196.64
Percent Purity 95%
Chemical Name or Material 3-(Chloromethyl)-5-(2-furyl)-1-methyl-1H-pyrazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.