accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 876728-37-9

IUPAC Name: 3-[(1H-1,2,4-triazol-1-yl)methyl]benzonitrile
Synonyms: 3-(1,2,4-triazol-1-ylmethyl)benzonitrile
Molecular Weight (g/mol): 184.2
Molecular Formula: C10H8N4
InChi Key: ZYQNZNVPCHZDLT-UHFFFAOYSA-N
SMILES: N#CC1=CC(CN2C=NC=N2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results