CAS RN 877-24-7
Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Organic Carbon Standard, 1000 ppm C, Ricca Chemical
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, ACS reagent, acidimetric standard
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.22 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, 99.99%, acidimetric standard
CAS: 877-24-7 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, 99+%
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.22 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Potassium hydrogen phthalate, 99%
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 MDL Number: MFCD00013070 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M Synonym: potassium hydrogen phthalate,potassium biphthalate,monopotassium phthalate,potassium acid phthalate,hydrogen potassium phthalate,phthalic acid monopotassium salt,1,2-benzenedicarboxylic acid, monopotassium salt,phthalic acid, monopotassium salt,phthalic acid potassium salt,potassium phthalate monobasic PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Carbon Standard, (Potassium Hydrogen Phthalate), Certified, 1000ppm ±10ppm Organic (1mL = 1mg C) , LabChem™
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M PubChem CID: 23676735 IUPAC Name: potassium;2-carboxybenzoate SMILES: C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Buffer Concentrate, pH 4.00 (Certified), Fisher Chemical™
Buffer Solution, pH 5.00 (Certified), Fisher Chemical™