accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 877399-00-3

IUPAC Name: 5-bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine
Synonyms: 5-bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine
Molecular Weight (g/mol): 380.04
Molecular Formula: C13H10BrCl2FN2O
InChi Key: URFUZAZEKBBCEY-UHFFFAOYNA-N
SMILES: CC(OC1=CC(Br)=CN=C1N)C1=C(Cl)C=CC(F)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results