Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine 98.0+%, TCI America™
SDP

Catalog No. B56825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56825G 5 g
B56821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56825G Supplier TCI America Supplier No. B56825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 877399-00-3
Molecular Formula C13H10BrCl2FN2O
Molecular Weight (g/mol) 380.038
MDL Number MFCD18207061
InChI Key URFUZAZEKBBCEY-ZCFIWIBFSA-N
PubChem CID 11689426
IUPAC Name 5-bromo-3-[(1R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine
SMILES CC(C1=C(C=CC(=C1Cl)F)Cl)OC2=CC(=CN=C2N)Br

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 5 g
Formula Weight 380.04
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-5-Bromo-3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]pyridin-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.