accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNBrFBFFF

CAS RN 878-23-9

IUPAC Name: 2-bromo-1-ethylpyridin-1-ium; tetrafluoroboranuide
Synonyms: 2-bromo-1-ethylpyridin-1-ium tetrafluoroborate
Molecular Weight (g/mol): 273.86
Molecular Formula: C7H9BBrF4N
InChi Key: YJDXVQLBIAJTHP-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC[N+]1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)