Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 97%

Catalog No. AC439960050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439960050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC439960050 Supplier Thermo Scientific Chemicals Supplier No. 439960050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 878-23-9 | C7H9BBrF4N | 273.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 878-23-9
Molecular Formula C7H9BBrF4N
Molecular Weight (g/mol) 273.86
MDL Number MFCD00060046
InChI Key YJDXVQLBIAJTHP-UHFFFAOYSA-N
Synonym 2-bromo-1-ethylpyridinium tetrafluoroborate, 2-bromo-1-ethylpyridin-1-ium tetrafluoroborate, bep tetrafluoroborate, bromoethylpyridiniumtetrafluoroborate, 2-bromo-1-ethyl-pyridinium tetrafluoroborate, bepybr, acmc-209qqc, 2-brom-1-ethyl-pyridiniumtetrafluoroborat, 2-bromo-1-ethyl pyridinium tetrafluoroborate, 2-bromo-1-ethyl-pyridin-1-ium tetrafluoroborate
PubChem CID 2736300
IUPAC Name 2-bromo-1-ethylpyridin-1-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC[N+]1=CC=CC=C1Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 100.0°C to 106.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 273.86
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1-ethyl-pyridinium tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 212-900-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.