accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 878376-35-3

IUPAC Name: pyridine-2-sulfonyl fluoride
Synonyms: pyridine-2-sulfonyl fluoride
Molecular Weight (g/mol): 161.15
Molecular Formula: C5H4FNO2S
InChi Key: FCFXLXGZHDHJLB-UHFFFAOYSA-N
SMILES: FS(=O)(=O)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)