Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-2-sulfonyl Fluoride 98.0+%, TCI America™
SDP

Catalog No. P23985G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23985G Supplier TCI America Supplier No. P23985G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 878376-35-3
Molecular Formula C5H4FNO2S
Molecular Weight (g/mol) 161.15
MDL Number MFCD09265513
InChI Key FCFXLXGZHDHJLB-UHFFFAOYSA-N
Synonym PyFluor
PubChem CID 11615218
IUPAC Name pyridine-2-sulfonyl fluoride
SMILES C1=CC=NC(=C1)S(=O)(=O)F

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 161.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Pyridine-2-sulfonyl Fluoride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.