accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHHOOPOOHOOHNa(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 87860-39-7

IUPAC Name: sodium 6-[4-(hydrogen phosphonatooxy)-3,6,13-trihydroxy-3-methyltrideca-1,7,9,11-tetraen-1-yl]-5,6-dihydro-2H-pyran-2-one
Synonyms: sodium 6-[4-(hydrogen phosphonatooxy)-3,6,13-trihydroxy-3-methyltrideca-1,7,9,11-tetraen-1-yl]-5,6-dihydropyran-2-one
Molecular Weight (g/mol): 452.37
Molecular Formula: C19H26NaO9P
InChi Key: XBUIKNRVGYFSHL-UHFFFAOYNA-M
SMILES: [Na+].CC(O)(C=CC1CC=CC(=O)O1)C(CC(O)C=CC=CC=CCO)OP(O)([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results