Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fostriecin sodium salt, Tocris Bioscience™
GSA_VA

Catalog No. 184050U
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-405-0U 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-405-0U Supplier R&D Systems Supplier No. 1840/50U

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Potent PP2A and PP4 inhibitor

Potent inhibitor of protein phosphatase types 2A (PP2A) and 4 (PP4) with IC50 values of 1.5 nM and 3 nM respectively. Also exhibits weaker inhibition of topoisomerase II (IC50 = 40 μM) and protein phosphatase type 1 (PP1) (IC50 = 131 μM) with no apparent inhibition of protein phosphatase type 2B (PP2B). Active in vivo. Originally identified as an antitumor antibiotic from Streptomyces pulveraceous.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 87860-39-7
Molecular Formula C19H26NaO9P
Molecular Weight (g/mol) 452.372
InChI Key XBUIKNRVGYFSHL-SOAGNPLSSA-M
Synonym fostriecin sodium salt
PubChem CID 124080892
IUPAC Name sodium;[(1Z,3R,4R,6R,7E,9E,11E)-3,6,13-trihydroxy-3-methyl-1-[(2R)-6-oxo-2,3-dihydropyran-2-yl]trideca-1,7,9,11-tetraen-4-yl] hydrogen phosphate
SMILES CC(C=CC1CC=CC(=O)O1)(C(CC(C=CC=CC=CCO)O)OP(=O)(O)[O-])O.[Na+]

Specifications

Quantity 50 μg
Formula Weight 452.37
Percent Purity >98%
Chemical Name or Material Fostriecin sodium salt
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.