accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 879-18-5

IUPAC Name: naphthalene-1-carbonyl chloride
Synonyms: naphthalene-1-carbonyl chloride
Molecular Weight (g/mol): 190.63
Molecular Formula: C11H7ClO
InChi Key: NSNPSJGHTQIXDO-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)