Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Naphthoyl chloride

Catalog No. 25025250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-025-250G 50 g
25-025-210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-025-250G Supplier Sigma Aldrich Supplier No. 25025250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Naphthoyl chloride

Specifications

CAS 879-18-5
Melting Point 16°C to 19°C (lit.)
Density 1.265 g/mL (at 25°C (literature))
Boiling Point 190°C (35 mmHg)
Molecular Formula C11H7ClO
Refractive Index n20/D 1.652 (literature)
Linear Formula C10H7COCl
MDL Number MFCD00004002
Quantity 50 g
Molecular Weight (g/mol) 190.63
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.