accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 879127-68-1

IUPAC Name: 4-fluoro-3,5-diiodobenzaldehyde
Synonyms: 4-fluoro-3,5-diiodobenzaldehyde
Molecular Weight (g/mol): 375.91
Molecular Formula: C7H3FI2O
InChi Key: QOHZTGBAXWWHTP-UHFFFAOYSA-N
SMILES: FC1=C(I)C=C(C=O)C=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results