accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOH

CAS RN 879360-14-2

IUPAC Name: decahydronaphthalene-1,4-dicarboxylic acid
Synonyms: decahydronaphthalene-1,4-dicarboxylic acid
Molecular Weight (g/mol): 226.27
Molecular Formula: C12H18O4
InChi Key: FEMXBCRRAJLQAS-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC(C2CCCCC12)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results