Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decahydro-1,4-naphthalenedicarboxylic Acid (mixture of isomers) 96.0+%, TCI America™
SDP

Catalog No. D43831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D43831G Supplier TCI America Supplier No. D43831G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 879360-14-2
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD27976820
InChI Key FEMXBCRRAJLQAS-UHFFFAOYNA-N
PubChem CID 18691683
IUPAC Name decahydronaphthalene-1,4-dicarboxylic acid
SMILES OC(=O)C1CCC(C2CCCCC12)C(O)=O

Specifications

Quantity 1 g
Formula Weight 226.27
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Decahydro-1,4-naphthalenedicarboxylic Acid (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.