accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 87981-04-2

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 11-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)undecanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 11-(2,5-dioxopyrrol-1-yl)undecanoate
Molecular Weight (g/mol): 378.43
Molecular Formula: C19H26N2O6
InChi Key: SGNVTRHWJGTNKV-UHFFFAOYSA-N
SMILES: O=C(CCCCCCCCCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results